accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COBr

CAS RN 68251-77-4

IUPAC Name: 2-bromo-3-methoxynaphthalene
Synonyms: 2-bromo-3-methoxynaphthalene
Molecular Weight (g/mol): 237.1
Molecular Formula: C11H9BrO
InChi Key: VFAPZPJQVMNTEX-UHFFFAOYSA-N
SMILES: COC1=C(Br)C=C2C=CC=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results