accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OBr

CAS RN 683-57-8

IUPAC Name: 2-bromoacetamide
Synonyms: bromoacetamide
Molecular Weight (g/mol): 137.96
Molecular Formula: C2H4BrNO
InChi Key: JUIKUQOUMZUFQT-UHFFFAOYSA-N
SMILES: NC(=O)CBr
Molecular Weight (g/mol) 
  • (4)
  • (4)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (4)
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (4)
  • (4)
  • (2)
  • (6)
  • (4)
  • (4)
  • (4)