accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3PClCl

CAS RN 683-85-2

IUPAC Name: (dichlorophosphanyl)dimethylamine
Synonyms: (dichlorophosphanyl)dimethylamine
Molecular Weight (g/mol): 145.95
Molecular Formula: C2H6Cl2NP
InChi Key: XPWWDZRSNFSLRQ-UHFFFAOYSA-N
SMILES: CN(C)P(Cl)Cl
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)