accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6830-78-0

NNNN

CAS RN 6830-78-0

IUPAC Name: diphenyl-1,2,4,5-tetrazine
Synonyms: diphenyl-1,2,4,5-tetrazine
Molecular Weight (g/mol): 234.26
Molecular Formula: C14H10N4
InChi Key: XAUWSIIGUUMHQQ-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=NN=C(N=N1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results