accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6830-83-7

IUPAC Name: N,2,2-trimethylpropanamide
Synonyms: N,2,2-trimethylpropanamide
Molecular Weight (g/mol): 115.18
Molecular Formula: C6H13NO
InChi Key: QMKKJBRRKIKWFK-UHFFFAOYSA-N
SMILES: CNC(=O)C(C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results