accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6832-87-7

IUPAC Name: 2-bromo-N-methylaniline
Synonyms: 2-bromo-N-methylaniline
Molecular Weight (g/mol): 186.05
Molecular Formula: C7H8BrN
InChi Key: SMVIAQFTVWDWDS-UHFFFAOYSA-N
SMILES: CNC1=CC=CC=C1Br
Boiling Point 
  • (4)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results