accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHN

CAS RN 6832-98-0

IUPAC Name: N-phenylhexadecanamide
Synonyms: N-phenylhexadecanamide
Molecular Weight (g/mol): 331.54
Molecular Formula: C22H37NO
InChi Key: VENJCACPSJGPCM-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC(=O)NC1=CC=CC=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)