accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHH2NHCl

CAS RN 68327-11-7

IUPAC Name: 2-aminocyclopentan-1-ol hydrochloride
Synonyms: 2-aminocyclopentan-1-ol hydrochloride
Molecular Weight (g/mol): 137.61
Molecular Formula: C5H12ClNO
InChi Key: ZFSXKSSWYSZPGQ-UHFFFAOYNA-N
SMILES: Cl.NC1CCCC1O
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)