accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClONH

CAS RN 6833-13-2

IUPAC Name: 2-chloro-N-phenylbenzamide
Synonyms: 2-chloro-N-phenylbenzamide
Molecular Weight (g/mol): 231.68
Molecular Formula: C13H10ClNO
InChi Key: AXQVIWHAEYLGLO-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1C(=O)NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results