accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3H3CCH3HNHOSSNSNH2O

CAS RN 68377-91-3

IUPAC Name: hydrogen 5-(2-{[3-(tert-butylamino)-2-hydroxypropyl]sulfanyl}-1,3-thiazol-4-yl)thiophene-2-carboxamide chloride
Synonyms: hydrogen arotinolol chloride
Molecular Weight (g/mol): 407.99
Molecular Formula: C15H22ClN3O2S3
InChi Key: XXDAXBZYUXLDRD-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(C)(C)NCC(O)CSC1=NC(=CS1)C1=CC=C(S1)C(N)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results