accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 68385-95-5

IUPAC Name: 2-amino-3,5-dibromobenzonitrile
Synonyms: 2-amino-3,5-dibromobenzonitrile
Molecular Weight (g/mol): 275.93
Molecular Formula: C7H4Br2N2
InChi Key: WLZCMGXAEXFAQA-UHFFFAOYSA-N
SMILES: NC1=C(Br)C=C(Br)C=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results