CAS RN 68392-35-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
3
4-Hydroxytamoxifen (E) and (Z) isomers (50:50), ≥97.5% (HPLC), MilliporeSigma™ Supelco™
4-Hydroxytamoxifen is a first generation, selective estrogen receptor modulator (SERM) that functions as an antagonist in breast cancer cells but can display estrogen-like activities in the uterus and bone.