CAS RN 68411-30-3
IUPAC Name:
ethane; sodium benzenesulfonate
Synonyms:
ethane; sodium benzene sulfonate
Molecular Weight (g/mol):
180.15
Molecular Formula:
C6H5NaO3S
InChi Key:
—
SMILES:
**.[Na]OS(=O)(=O)C1=CC=CC=C1
1
–
2
of
2
results
2
Linear Alkylbenzene Sulfonate Standard, 1000 ppm LAS, Ricca Chemical
CAS: 68411-30-3 Molecular Formula: C6H5NaO3S Molecular Weight (g/mol): 180.15