accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6843-66-9

H3COSiOCH3

CAS RN 6843-66-9

IUPAC Name: dimethoxydiphenylsilane
Synonyms: silane, dimethoxydiphenyl-
Molecular Weight (g/mol): 244.37
Molecular Formula: C14H16O2Si
InChi Key: AHUXYBVKTIBBJW-UHFFFAOYSA-N
SMILES: CO[Si](OC)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)