accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 685-83-6

IUPAC Name: [chloro(diethylamino)phosphanyl]diethylamine
Synonyms: [chloro(diethylamino)phosphanyl]diethylamine
Molecular Weight (g/mol): 210.69
Molecular Formula: C8H20ClN2P
InChi Key: JVEHJSIFWIIFHM-UHFFFAOYSA-N
SMILES: CCN(CC)P(Cl)N(CC)CC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)