accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOBrOOCH3

CAS RN 685-87-0

IUPAC Name: 1,3-diethyl 2-bromopropanedioate
Synonyms: 1,3-diethyl 2-bromopropanedioate
Molecular Weight (g/mol): 239.07
Molecular Formula: C7H11BrO4
InChi Key: FNJVDWXUKLTFFL-UHFFFAOYSA-N
SMILES: CCOC(=O)C(Br)C(=O)OCC
Boiling Point 
  • (6)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (6)
  • (2)
  • (2)
  • (2)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (5)