accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 685-91-6

IUPAC Name: N,N-diethylacetamide
Synonyms: diethylacetamide
Molecular Weight (g/mol): 115.18
Molecular Formula: C6H13NO
InChi Key: AJFDBNQQDYLMJN-UHFFFAOYSA-N
SMILES: CCN(CC)C(C)=O
Boiling Point 
  • (3)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (5)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)