accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONH3HOOHHO

CAS RN 6850-28-8

IUPAC Name: 1,3-dihydroxy-2-(hydroxymethyl)propan-2-aminium acetate
Synonyms: htris acetate
Molecular Weight (g/mol): 181.19
Molecular Formula: C6H15NO5
InChi Key: PIEPQKCYPFFYMG-UHFFFAOYSA-N
SMILES: CC([O-])=O.[NH3+]C(CO)(CO)CO
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (2)
  • (1)
  • (2)