accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 68504-35-8

IUPAC Name: tetrasodium (3Z,6Z)-4,5-dioxo-3,6-bis[2-(2-sulfonatophenyl)hydrazin-1-ylidene]-3,4,5,6-tetrahydronaphthalene-2,7-disulfonate
Synonyms: tetrasodium (3Z,6Z)-4,5-dioxo-3,6-bis[2-(2-sulfonatophenyl)hydrazin-1-ylidene]naphthalene-2,7-disulfonate
Molecular Weight (g/mol): 776.55
Molecular Formula: C22H12N4Na4O14S4
InChi Key: OWVXGCUJHKOBMQ-DTSPTRIMSA-J
SMILES: [Na+].[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=CC=C1N\N=C1\C(=O)C2=C(C=C1S([O-])(=O)=O)C=C(\C(=N/NC1=CC=CC=C1S([O-])(=O)=O)C2=O)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)