accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3CH3H3CCH3OOCH3H2C

CAS RN 68548-08-3

IUPAC Name: 1,2,2,6,6-pentamethylpiperidin-4-yl 2-methylprop-2-enoate
Synonyms: 1,2,2,6,6-pentamethylpiperidin-4-yl 2-methylprop-2-enoate
Molecular Weight (g/mol): 239.36
Molecular Formula: C14H25NO2
InChi Key: NWPIOULNZLJZHU-UHFFFAOYSA-N
SMILES: CN1C(C)(C)CC(CC1(C)C)OC(=O)C(C)=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results