accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH(S)H2N

CAS RN 6859-99-0

IUPAC Name: (3S)-3-hydroxypiperidin-1-ium
Synonyms: (3S)-3-hydroxypiperidin-1-ium
Molecular Weight (g/mol): 102.16
Molecular Formula: C5H12NO
InChi Key: BIWOSRSKDCZIFM-YFKPBYRVSA-O
SMILES: O[C@H]1CCC[NH2+]C1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (3)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)