accessibility menu, dialog, popup

UserName

Viewing profile as user:
OH(R)O(S)O(R)HO(S)OH(R)OH(R)OH(R)OHO

CAS RN 6860-47-5

IUPAC Name: (2R,3R,4R)-2,3,5-trihydroxy-4-{[(2S,3R,4S,5R)-3,4,5-trihydroxyoxan-2-yl]oxy}pentanal
Synonyms: xylobiose
Molecular Weight (g/mol): 282.25
Molecular Formula: C10H18O9
InChi Key: SQNRKWHRVIAKLP-RSZZQXBVSA-N
SMILES: OC[C@@H](O[C@@H]1OC[C@@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)