accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OONHH2N

CAS RN 68621-88-5

IUPAC Name: tert-butyl N-(3-aminophenyl)carbamate
Synonyms: tert-butyl N-(3-aminophenyl)carbamate
Molecular Weight (g/mol): 208.26
Molecular Formula: C11H16N2O2
InChi Key: IEUIEMIRUXSXCL-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC=CC(N)=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)