accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6864-20-6

IUPAC Name: 3,5-diphenyl-[1,1'-biphenyl]-2-amine
Synonyms: 3,5-diphenyl-[1,1'-biphenyl]-2-amine
Molecular Weight (g/mol): 321.42
Molecular Formula: C24H19N
InChi Key: AGWKGKUGJDMKMT-UHFFFAOYSA-N
SMILES: NC1=C(C=C(C=C1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)