accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 68641-49-6

IUPAC Name: bis(2-oxo-1,3-oxazolidin-3-yl)phosphinoyl chloride
Synonyms: bis(2-oxo-1,3-oxazolidin-3-yl)phosphinoyl chloride
Molecular Weight (g/mol): 254.56
Molecular Formula: C6H8ClN2O5P
InChi Key: KLDLRDSRCMJKGM-UHFFFAOYSA-N
SMILES: ClP(=O)(N1CCOC1=O)N1CCOC1=O
Molecular Weight (g/mol) 
  • (2)
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (4)
Quantity 
  • (4)
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (4)
  • (4)
  • (4)