accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6867-30-7

IUPAC Name: ethane-1,2-diamine; ethynyllithium
Synonyms: ethylenediamine; ethynyllithium
Molecular Weight (g/mol): 92.07
Molecular Formula: C4H9LiN2
InChi Key: QJQWXTYPTBEPGS-UHFFFAOYSA-N
SMILES: [Li]C#C.NCCN
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (5)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (5)
  • (5)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)
  • (2)
  • (2)
  • (2)