accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2HOONHOHO

CAS RN 687-63-8

IUPAC Name: 2-(2-amino-3-hydroxypropanamido)acetic acid
Synonyms: (2-amino-3-hydroxypropanamido)acetic acid
Molecular Weight (g/mol): 162.15
Molecular Formula: C5H10N2O4
InChi Key: WOUIMBGNEUWXQG-UHFFFAOYNA-N
SMILES: NC(CO)C(=O)NCC(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results