accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 68737-65-5

IUPAC Name: (1R,2R)-N1,N2-dimethylcyclohexane-1,2-bis(aminium)
Synonyms: (1R,2R)-N1,N2-dimethylcyclohexane-1,2-bis(aminium)
Molecular Weight (g/mol): 144.26
Molecular Formula: C8H20N2
InChi Key: JRHPOFJADXHYBR-HTQZYQBOSA-P
SMILES: C[NH2+][C@@H]1CCCC[C@H]1[NH2+]C
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)