accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrO

CAS RN 6876-00-2

IUPAC Name: 1-bromo-3-phenoxybenzene
Synonyms: 1-bromo-3-phenoxybenzene
Molecular Weight (g/mol): 249.11
Molecular Formula: C12H9BrO
InChi Key: AHDAKFFMKLQPTD-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(OC2=CC=CC=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results