CAS RN 6876-23-9
IUPAC Name:
(1S,2S)-1,2-dimethylcyclohexane
Synonyms:
(1S,2S)-1,2-dimethylcyclohexane
Molecular Weight (g/mol):
112.22
Molecular Formula:
C8H16
InChi Key:
KVZJLSYJROEPSQ-YUMQZZPRSA-N
SMILES:
C[C@H]1CCCC[C@@H]1C
1
–
1
of
1
results
1
trans-1,2-Dimethylcyclohexane 99.0+%, TCI America™
CAS: 6876-23-9 Molecular Formula: C8H16 Molecular Weight (g/mol): 112.216 MDL Number: MFCD00001505 InChI Key: KVZJLSYJROEPSQ-YUMQZZPRSA-N Synonym: trans-Hexahydro-o-xylene PubChem CID: 252360 IUPAC Name: (1S,2S)-1,2-dimethylcyclohexane SMILES: CC1CCCCC1C