CAS RN 68767-14-6
CAS RN 68767-14-6
IUPAC Name:
2-{4-[(2-oxocyclopentyl)methyl]phenyl}propanoic acid
Synonyms:
loxoprofen 洛索洛芬
Molecular Weight (g/mol):
246.31
Molecular Formula:
C15H18O3
InChi Key:
YMBXTVYHTMGZDW-UHFFFAOYNA-N
SMILES:
CC(C(O)=O)C1=CC=C(CC2CCCC2=O)C=C1
1
–
1
of
1
results
1
Loxoprofen 97.0+%, TCI America™

CAS: 68767-14-6 Molecular Formula: C15H18O3 Molecular Weight (g/mol): 246.31 MDL Number: MFCD00864331 InChI Key: YMBXTVYHTMGZDW-UHFFFAOYNA-N Synonym: 2-[4-[(2-Oxocyclopentyl)methyl]phenyl]propionic Acid PubChem CID: 3965 ChEBI: CHEBI:76172 IUPAC Name: 2-{4-[(2-oxocyclopentyl)methyl]phenyl}propanoic acid SMILES: CC(C(O)=O)C1=CC=C(CC2CCCC2=O)C=C1