accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 68786-66-3

IUPAC Name: 6-chloro-5-(2,3-dichlorophenoxy)-2-(methylsulfanyl)-1H-1,3-benzodiazole
Synonyms: triclabendazole triclabendazol C14H9cl3N2OS flukare virbac brand of triclabendazole triclabendazolum jurox brand of triclabendazole fasinex tremacide novartis brand of triclabendazole
Molecular Weight (g/mol): 359.65
Molecular Formula: C14H9Cl3N2OS
InChi Key: NQPDXQQQCQDHHW-UHFFFAOYSA-N
SMILES: CSC1=NC2=CC(OC3=CC=CC(Cl)=C3Cl)=C(Cl)C=C2N1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)