accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 688-74-4

IUPAC Name: tributyl borate
Synonyms: butyl borate
Molecular Weight (g/mol): 230.16
Molecular Formula: C12H27BO3
InChi Key: LGQXXHMEBUOXRP-UHFFFAOYSA-N
SMILES: CCCCOB(OCCCC)OCCCC
Boiling Point 
  • (2)
  • (3)
  • (3)
  • (4)
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (7)
Quantity 
  • (4)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
Percent Purity 
  • (3)
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (7)
  • (2)
  • (3)
  • (3)
  • (4)
  • (3)
  • (2)
  • (7)
  • (4)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (3)
  • (7)
  • (7)