accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6881-57-8

IUPAC Name: benzylphosphonate
Synonyms: benzylphosphonate
Molecular Weight (g/mol): 170.11
Molecular Formula: C7H7O3P
InChi Key: OGBVRMYSNSKIEF-UHFFFAOYSA-L
SMILES: [O-]P([O-])(=O)CC1=CC=CC=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results