CAS RN 68892-42-2
IUPAC Name:
N-{9-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6-oxo-6,9-dihydro-3H-purin-2-yl}-2-methylpropanamide
Synonyms:
N-{9-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6-oxo-3H-purin-2-yl}-2-methylpropanamide
Molecular Weight (g/mol):
337.34
Molecular Formula:
C14H19N5O5
InChi Key:
SIDXEQFMTMICKG-UHFFFAOYNA-N
SMILES:
CC(C)C(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C1CC(O)C(CO)O1
1
–
1
of
1
results
1
N2-Isobutyryl-2'-deoxyguanosine 98.0+%, TCI America™
CAS: 68892-42-2 Molecular Formula: C14H19N5O5 Molecular Weight (g/mol): 337.336 MDL Number: MFCD00010060 InChI Key: SIDXEQFMTMICKG-DJLDLDEBSA-N PubChem CID: 688078 IUPAC Name: N-[9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6-oxo-3H-purin-2-yl]-2-methylpropanamide SMILES: CC(C)C(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C3CC(C(O3)CO)O