accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 68931-30-6

IUPAC Name: 3,7,11,15-tetramethylhexadeca-1,6,10,14-tetraen-3-ol
Synonyms: 3,7,11,15-tetramethylhexadeca-1,6,10,14-tetraen-3-ol
Molecular Weight (g/mol): 290.49
Molecular Formula: C20H34O
InChi Key: IQDXAJNQKSIPGB-UHFFFAOYNA-N
SMILES: CC(C)=CCCC(C)=CCCC(C)=CCCC(C)(O)C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results