CAS RN 68938-92-1
IUPAC Name:
tetrabromoplatinumtetrakis(ylium)
Synonyms:
tetrabromoplatinumtetrakis(ylium)
Molecular Weight (g/mol):
514.7
Molecular Formula:
Br4Pt
InChi Key:
DYFRBYUUSCNRBP-UHFFFAOYSA-J
SMILES:
Br[Pt+4](Br)(Br)Br
1
–
1
of
1
results
1
Platinum(IV) bromide, 99.99% (metals basis), Pt 37.1% min, Thermo Scientific Chemicals
CAS: 68938-92-1 Molecular Formula: Br4Pt Molecular Weight (g/mol): 514.70 MDL Number: MFCD00016289 InChI Key: DYFRBYUUSCNRBP-UHFFFAOYSA-J Synonym: platinum tetrabromide,platinum iv bromide,platinum 4+ tetrabromide IUPAC Name: tetrabromoplatinumtetrakis(ylium) SMILES: Br[Pt+4](Br)(Br)Br