accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 68959-06-8

IUPAC Name: bis(butan-2-yloxy)alumanyl triethyl silicate
Synonyms: bis(sec-butoxy)alumanyl triethyl silicate
Molecular Weight (g/mol): 352.48
Molecular Formula: C14H33AlO6Si
InChi Key: MTHHTRVJDXYJHE-UHFFFAOYNA-N
SMILES: CCO[Si](OCC)(OCC)O[Al](OC(C)CC)OC(C)CC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results