accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 68959-87-5

IUPAC Name: 2,2',2''-tris(propan-2-yl)-1-lanthana-1,1',1''-spiroter[pentacyclo[2.2.0.0¹,³.0¹,⁵.0²,⁶]hexane]
Synonyms: 2,2',2''-triisopropyl-1-lanthana-1,1',1''-spiroter[pentacyclo[2.2.0.0¹,³.0¹,⁵.0²,⁶]hexane]
Molecular Weight (g/mol): 460.43
Molecular Formula: C24H33La
InChi Key: BHWFVSOAOOHARF-UHFFFAOYNA-N
SMILES: CC(C)C12[La]3456789%10%11%12%13%14%15C(C3C14)C25.CC(C)C61C7C8C9C%101.CC(C)C%111C%12C%13C%14C%151

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results