accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 68981-86-2

IUPAC Name: 3-(4,4-dimethyl-4,5-dihydro-1,3-oxazol-2-yl)pyridine
Synonyms: 3-(4,4-dimethyl-5H-1,3-oxazol-2-yl)pyridine
Molecular Weight (g/mol): 176.22
Molecular Formula: C10H12N2O
InChi Key: XYABHURTMBEOGW-UHFFFAOYSA-N
SMILES: CC1(C)COC(=N1)C1=CN=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results