accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6899-04-3

IUPAC Name: 2-amino-4-carbamoylbutanoic acid
Synonyms: glutamin Hgln glutamic acid gamma-amide glutaminsaeure-5-amid
Molecular Weight (g/mol): 146.15
Molecular Formula: C5H10N2O3
InChi Key: ZDXPYRJPNDTMRX-UHFFFAOYNA-N
SMILES: NC(CCC(N)=O)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)