accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 69-78-3

IUPAC Name: 5-[(3-carboxy-4-nitrophenyl)disulfanyl]-2-nitrobenzoic acid
Synonyms: ellman's reagent dithionitrobenzoic acid
Molecular Weight (g/mol): 396.34
Molecular Formula: C14H8N2O8S2
InChi Key: KIUMMUBSPKGMOY-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(SSC2=CC=C(C(=C2)C(O)=O)[N+]([O-])=O)=CC=C1[N+]([O-])=O
Percent Purity 
  • (1)
  • (3)
  • (6)
Quantity 
  • (3)
  • (3)
  • (4)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (3)
  • (6)
  • (1)
  • (3)
  • (6)
  • (3)
  • (3)
  • (4)
  • (3)
  • (1)