CAS RN 690-37-9
IUPAC Name:
2,4,4-trimethylpentan-2-ol
Synonyms:
2,4,4-trimethylpentan-2-ol
Molecular Weight (g/mol):
130.23
Molecular Formula:
C8H18O
InChi Key:
BSYJHYLAMMJNRC-UHFFFAOYSA-N
SMILES:
CC(C)(C)CC(C)(C)O
1
–
1
of
1
results
1
2,4,4-Trimethyl-2-pentanol 95.0+%, TCI America™
CAS: 690-37-9 Molecular Formula: C8H18O Molecular Weight (g/mol): 130.231 MDL Number: MFCD00101611 InChI Key: BSYJHYLAMMJNRC-UHFFFAOYSA-N Synonym: 2-Hydroxy-2,4,4-trimethylpentane PubChem CID: 94740 IUPAC Name: 2,4,4-trimethylpentan-2-ol SMILES: CC(C)(C)CC(C)(C)O