accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 690206-97-4

IUPAC Name: N-(4-chloro-2-fluorophenyl)-6,7-dimethoxyquinazolin-4-amine
Synonyms: N-(4-chloro-2-fluorophenyl)-6,7-dimethoxyquinazolin-4-amine
Molecular Weight (g/mol): 333.75
Molecular Formula: C16H13ClFN3O2
InChi Key: YHUIUSRCUKUUQA-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C2C(NC3=C(F)C=C(Cl)C=C3)=NC=NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results