accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 69026-14-8

IUPAC Name: 3-(benzyloxy)benzoate
Synonyms: 3-(benzyloxy)benzoate
Molecular Weight (g/mol): 227.24
Molecular Formula: C14H11O3
InChi Key: CISXCTKEQYOZAM-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=CC(OCC2=CC=CC=C2)=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results