accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClIN

CAS RN 69045-79-0

IUPAC Name: 2-chloro-5-iodopyridine
Synonyms: 2-chloro-5-iodopyridine
Molecular Weight (g/mol): 239.44
Molecular Formula: C5H3ClIN
InChi Key: QWLGCWXSNYKKDO-UHFFFAOYSA-N
SMILES: ClC1=CC=C(I)C=N1
Percent Purity 
  • (2)
  • (1)
  • (3)
  • (3)
Quantity 
  • (3)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (6)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)