accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 690632-17-8

BOOCH3H3CCH3CH3NSiH3CCH3CH3CH3H3CH3C

CAS RN 690632-17-8

IUPAC Name: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-[tris(propan-2-yl)silyl]-1H-indole
Synonyms: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-(triisopropylsilyl)indole
Molecular Weight (g/mol): 399.46
Molecular Formula: C23H38BNO2Si
InChi Key: NGMKDRPSWHSLIM-UHFFFAOYSA-N
SMILES: CC(C)[Si](C(C)C)(C(C)C)N1C=CC2=C(C=CC=C12)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results