accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 690632-88-3

IUPAC Name: 4-methyl-2-phenyl-1,3-thiazole-5-sulfonyl chloride
Synonyms: 4-methyl-2-phenyl-1,3-thiazole-5-sulfonyl chloride
Molecular Weight (g/mol): 273.75
Molecular Formula: C10H8ClNO2S2
InChi Key: NGDQQLAVJWUYSF-UHFFFAOYSA-N
SMILES: CC1=C(SC(=N1)C1=CC=CC=C1)S(Cl)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results