accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 69064-37-5

IUPAC Name: dodec-2-en-1-ol
Synonyms: 2-dodecenol
Molecular Weight (g/mol): 184.32
Molecular Formula: C12H24O
InChi Key: MLRYPOCSLBIUHY-UHFFFAOYSA-N
SMILES: CCCCCCCCCC=CCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results