accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COHH2N

CAS RN 69088-96-6

IUPAC Name: 4-(3-aminophenyl)-2-methylbut-3-yn-2-ol
Synonyms: 4-(3-aminophenyl)-2-methylbut-3-yn-2-ol
Molecular Weight (g/mol): 175.23
Molecular Formula: C11H13NO
InChi Key: DQPSETABKZMTEZ-UHFFFAOYSA-N
SMILES: CC(C)(O)C#CC1=CC=CC(N)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results